Orders LinkContactUsLinkComponent
Taurine ≥99%
Glass bottle for solids, 500 g
  A12403.36
Taurine ≥99%
  A12403.36
 :  107-35-7
Taurine ≥99%
Taurine ≥99%
Glass bottle for solids, 500 g
 :  107-35-7
99,40 €
1 * 500 g
Purity: 99% CAS No.: 107-35-7 Molecular Formula: H2NCH2CH2SO3H Formula Weight: 125.15 Physical Form: Crystals or cryst powder Appearance: White Melting Point: 300° Beilstein Reference: 1751215 Merck Reference: 14,9074 MDL No.: MFCD00008197
 :  2-Aminoethanesulphonic acid
  •  :107-35-7
  •  :C₂H₇NO₃S
  •  :MFCD00008197
  •  :125.15 g/mol
  •  :14,9074
  •  :>300 °C
  •  :Ambient
  •  :203-483-8
  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g