Order Entry
Export
Orders LinkContactUsLinkComponent
Taurine ≥99%

Taurine ≥99%

Glass bottle for solids, 100 g
Catalog # A12403.22
Irritant
Taurine ≥99%
Catalog # A12403.22
CAS Number:  107-35-7
Taurine ≥99%
Taurine ≥99%

Glass bottle for solids, 100 g

CAS Number:  107-35-7
Price
36,20 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 99% CAS No.: 107-35-7 Molecular Formula: H2NCH2CH2SO3H Formula Weight: 125.15 Physical Form: Crystals or cryst powder Appearance: White Melting Point: 300° Beilstein Reference: 1751215 Merck Reference: 14,9074 MDL No.: MFCD00008197

Synonyms:  2-Aminoethanesulphonic acid
  • CAS Number:107-35-7
  • Formula:C₂H₇NO₃S
  • MDL Number:MFCD00008197
  • Molecular Weight:125.15 g/mol
  • Merck Index:14,9074
  • Melting Pt:>300 °C
  • Storage Temperature:Ambient
  • EINECS:203-483-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications