Urea ≥98%
Glass bottle for solids, 2.500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 98+% CAS No.: 57-13-6 Molecular Formula: NH2CONH2 Formula Weight: 60.06 Odour: Ammonia-like pH: 9,2-9,5 Melting Point: 130-135° Beilstein Reference: 635724 Merck Reference: 14,9867 MDL No.: MFCD00008022
Synonyms: Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide
- CAS Number:57-13-6
- Formula:NH₂CONH₂
- MDL Number:MFCD00008022
- Molecular Weight:60.06 g/mol
- Merck Index:14,9867
- Boiling Pt:197 °C (1013 hPa)
- Melting Pt:130…135 °C
- Density:1.335
- Storage Temperature:Ambient
- EINECS:200-315-5
- REACH:01-2119463277-33
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:2.500 g



