2,2'-Bithiophene ≥98%
Glass bottle for solids, 1 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 98% CAS No.: 492-97-7 Molecular Formula: C8H6S2 Formula Weight: 166.27 Flash Point: 110°(230°F) Melting Point: 30-33° Boiling Point: 260° Beilstein Reference: 3039 MDL No.: MFCD00005414
Synonyms: 2,2′-Dithienyl, 2,2′-Bithienyl
- CAS Number:492-97-7
- Formula:C₈H₆S₂
- MDL Number:MFCD00005414
- Molecular Weight:166.27 g/mol
- Boiling Pt:260 °C
- Melting Pt:30…33 °C
- Flash Pt:>110 °C (230 °F)
- Storage Temperature:Ambient
- EINECS:207-767-2
Documents
Certificates
Search by Lot NumberOrganic Building Blocks and Intermediates
Application Notes
Thermo Scientific Organic Photovoltaics
Product Literature
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



