Order Entry
Export
Orders LinkContactUsLinkComponent
3-Methylpyrocatechol ≥98%

3-Methylpyrocatechol ≥98%

Glass bottle for solids, 100 g
Catalog # A12324.22
Toxic
Irritant
3-Methylpyrocatechol ≥98%
Catalog # A12324.22
CAS Number:  488-17-5
3-Methylpyrocatechol ≥98%
3-Methylpyrocatechol ≥98%

Glass bottle for solids, 100 g

CAS Number:  488-17-5
Price
431,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 98% CAS No.: 488-17-5 Molecular Formula: C7H8O2 Formula Weight: 124.14 Flash Point: 140°(284°F) Melting Point: 62-68° Boiling Point: 241° Beilstein Reference: 774602 MDL No.: MFCD00016435

Synonyms:  3-Methylcatechol, 3-Methylbenzene-1,2-diol
  • CAS Number:488-17-5
  • Formula:C₇H₈O₂
  • MDL Number:MFCD00016435
  • Molecular Weight:124.14 g/mol
  • Boiling Pt:241 °C
  • Melting Pt:62…68 °C
  • Flash Pt:140 °C (284 °F)
  • Storage Temperature:Ambient
  • EINECS:207-672-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications