Order Entry
Export
Orders LinkContactUsLinkComponent
 

2-Bromotoluene ≥99%

Glass bottle for solids, 500 g
Catalog # A12315.36
Irritant
2-Bromotoluene ≥99%
Catalog # A12315.36
CAS Number:  95-46-5
2-Bromotoluene ≥99%
2-Bromotoluene ≥99%

Glass bottle for solids, 500 g

CAS Number:  95-46-5
Price
328,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 99% CAS No.: 95-46-5 Molecular Formula: C7H7Br Formula Weight: 171.04 Flash Point: 78°(172°F) Refractive Index: 1.5560 Melting Point: -27° Boiling Point: 179-181° Beilstein Reference: 1904176 Merck Reference: 14,1439 MDL No.: MFCD00000068

  • CAS Number:95-46-5
  • Formula:C₇H₇Br
  • MDL Number:MFCD00000068
  • Molecular Weight:171.04 g/mol
  • Merck Index:14,1439
  • Boiling Pt:179…181 °C
  • Melting Pt:–27 °C
  • Density:1.420
  • Flash Pt:78 °C (172 °F)
  • Storage Temperature:Ambient
  • EINECS:202-421-7
  • UN:3082
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications