DL-Alanine ≥99%
Glass bottle for solids, 250 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 302-72-7 Molecular Formula: C3H7NO2 Formula Weight: 89.09 Melting Point: ca 292° dec. Beilstein Reference: 635807 Merck Reference: 14,204 MDL No.: MFCD00064408
Synonyms: (±)-Alanine, DL-α-Alanine, H-DL-Ala-OH, α-Alanine
- CAS Number:302-72-7
- Formula:C₃H₇NO₂
- MDL Number:MFCD00064408
- Molecular Weight:89.09 g/mol
- Merck Index:14,204
- Boiling Pt:189 °C (1013 hPa)
- Melting Pt:∼ 292 °C (dec.)
- Density:1.42
- Storage Temperature:Ambient
- EINECS:206-126-4
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g



