Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,6-Dimethoxybenzaldehyde ≥99%

Glass bottle for solids, 25 g
Catalog # A12209.14
Irritant
2,6-Dimethoxybenzaldehyde ≥99%
Catalog # A12209.14
CAS Number:  3392-97-0
2,6-Dimethoxybenzaldehyde ≥99%
2,6-Dimethoxybenzaldehyde ≥99%

Glass bottle for solids, 25 g

CAS Number:  3392-97-0
Price
649,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 99% CAS No.: 3392-97-0 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 96-98° Boiling Point: 285° MDL No.: MFCD00010862

  • CAS Number:3392-97-0
  • Formula:C₉H₁₀O₃
  • MDL Number:MFCD00010862
  • Molecular Weight:166.18 g/mol
  • Boiling Pt:285 °C
  • Melting Pt:96…98 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications