Order Entry
Export
Orders LinkContactUsLinkComponent
 

1-Octadecanol ≥97%

Glass bottle for solids, 250 g
Catalog # A12020.30
1-Octadecanol ≥97%
Catalog # A12020.30
CAS Number:  112-92-5
1-Octadecanol ≥97%
1-Octadecanol ≥97%

Glass bottle for solids, 250 g

CAS Number:  112-92-5
Price
27,30 €
Log in for availability
1 * 250 g
Product Details & Documents

Purity: 97% CAS No.: 112-92-5 Molecular Formula: CH3(CH2)17OH Formula Weight: 270.50 Flash Point: 174°(345°F) Physical Form: Flakes or pellets Melting Point: 56-60° Boiling Point: 210°/15mm Beilstein Reference: 1362907 Merck Reference: 14,8805 MDL No.: MFCD00002823

Synonyms:  Stearyl alcohol, 1-Hydroxyoctadecane, Heptadecyl carbinol, 1-Octadecyl alcohol
  • CAS Number:112-92-5
  • Formula:H₃C(CH₂)₁₆CH₂OH
  • MDL Number:MFCD00002823
  • Molecular Weight:270.5 g/mol
  • Merck Index:14,8805
  • Boiling Pt:210 °C (15 mmHg)
  • Melting Pt:56…60 °C
  • Density:0.805…0.815 g/cm³ (60 °C)
  • Flash Pt:174 °C (345 °F)
  • Storage Temperature:Ambient
  • EINECS:204-017-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications