Orders LinkContactUsLinkComponent
2,5-Dimethyl-2,5-hexanediol ≥97%
2,5-Dimethyl-2,5-hexanediol ≥97%
Glass bottle for solids, 100 g
  A11984.22
2,5-Dimethyl-2,5-hexanediol ≥97%
  A11984.22
 :  110-03-2
2,5-Dimethyl-2,5-hexanediol ≥97%
2,5-Dimethyl-2,5-hexanediol ≥97%
Glass bottle for solids, 100 g
 :  110-03-2
29,80 €
1 * 100 g
Purity: 97% CAS No.: 110-03-2 Molecular Formula: C8H18O2 Formula Weight: 146.23 Flash Point: 126°(258°F) Odour: Odourless Boiling Point: 214-215° Beilstein Reference: 1361437 MDL No.: MFCD00004473
 :  1,1,4,4-Tetramethyl-1,4-butanediol
  •  :110-03-2
  •  :C₈H₁₈O₂
  •  :MFCD00004473
  •  :146.23 g/mol
  •  :214…215 °C
  •  :87…91 °C
  •  :0.898
  •  :126 °C (258 °F)
  •  :Ambient
  •  :203-731-5
  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g