Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,5-Dimethyl-2,5-hexanediol ≥97%

Glass bottle for solids, 2.500 g
Catalog # A11984.0E
Irritant
2,5-Dimethyl-2,5-hexanediol ≥97%
Catalog # A11984.0E
CAS Number:  110-03-2
2,5-Dimethyl-2,5-hexanediol ≥97%
2,5-Dimethyl-2,5-hexanediol ≥97%

Glass bottle for solids, 2.500 g

CAS Number:  110-03-2
Price
354,00 €
Log in for availability
1 * 2.500 g
Product Details & Documents

Purity: 97% CAS No.: 110-03-2 Molecular Formula: C8H18O2 Formula Weight: 146.23 Flash Point: 126°(258°F) Odour: Odourless Boiling Point: 214-215° Beilstein Reference: 1361437 MDL No.: MFCD00004473

Synonyms:  1,1,4,4-Tetramethyl-1,4-butanediol
  • CAS Number:110-03-2
  • Formula:C₈H₁₈O₂
  • MDL Number:MFCD00004473
  • Molecular Weight:146.23 g/mol
  • Boiling Pt:214…215 °C
  • Melting Pt:87…91 °C
  • Density:0.898
  • Flash Pt:126 °C (258 °F)
  • Storage Temperature:Ambient
  • EINECS:203-731-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2.500 g

Specifications