Order Entry
Export
Orders LinkContactUsLinkComponent
2-Methylresorcinol 98%

2-Methylresorcinol 98%

Glass bottle for solids, 1.000 g
Catalog # A11821.0B
Toxic
Irritant
2-Methylresorcinol 98%
Catalog # A11821.0B
CAS Number:  608-25-3
2-Methylresorcinol 98%
2-Methylresorcinol 98%

Glass bottle for solids, 1.000 g

CAS Number:  608-25-3
Price
588,00 €
Log in for availability
1 * 1.000 g
Product Details & Documents

Purity: 98% CAS No.: 608-25-3 Molecular Formula: C7H8O2 Formula Weight: 124.14 Flash Point: 135°(275°F) Melting Point: 118-122° Boiling Point: 264° Beilstein Reference: 2042177 UN No.: 2811 MDL No.: MFCD00002271

Synonyms:  2,6-Dihydroxytoluene, 2-Methylbenzene-1,3-diol
  • CAS Number:608-25-3
  • Formula:C₇H₈O₂
  • MDL Number:MFCD00002271
  • Molecular Weight:124.14 g/mol
  • Boiling Pt:264 °C
  • Melting Pt:118…122 °C
  • Flash Pt:135 °C (275 °F)
  • Storage Temperature:Ambient
  • EINECS:210-155-8
  • UN:2811
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1.000 g

Specifications