Order Entry
Export
Orders LinkContactUsLinkComponent
(±)-1,3-Butanediol ≥99%

(±)-1,3-Butanediol ≥99%

Glass bottle, 10.000 mL
Catalog # A11684.0D
(±)-1,3-Butanediol ≥99%
Catalog # A11684.0D
CAS Number:  107-88-0
(±)-1,3-Butanediol ≥99%
(±)-1,3-Butanediol ≥99%

Glass bottle, 10.000 mL

CAS Number:  107-88-0
Price
241,00 €
Log in for availability
1 * 10.000 mL
Product Details & Documents

Purity: 99% CAS No.: 107-88-0 Molecular Formula: C4H10O2 Formula Weight: 90.12 Flash Point: 109°(249°F) Refractive Index: 1.4400 Odour: Mild Melting Point: -50° Boiling Point: 203-204° Beilstein Reference: 1731276 Merck Reference: 14,1567 MDL No.: MFCD00004554

Synonyms:  Butane-1,3-diol, (±)-1,3-Butylene glycol, (±)-1,3-Dihydroxy butane
  • CAS Number:107-88-0
  • Formula:C₄H₁₀O₂
  • MDL Number:MFCD00004554
  • Molecular Weight:90.12 g/mol
  • Merck Index:14,1567
  • Boiling Pt:203…204 °C
  • Melting Pt:–50 °C
  • Density:1.005
  • Flash Pt:109 °C (249 °F)
  • Storage Temperature:Ambient
  • EINECS:203-529-7
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10.000 mL

Specifications