Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,4,6-Trichlorophenol ≥98%

Glass bottle for solids, 250 g
Catalog # A11681.30
Health
Irritant
Environment
2,4,6-Trichlorophenol ≥98%
Catalog # A11681.30
CAS Number:  88-06-2
2,4,6-Trichlorophenol ≥98%
2,4,6-Trichlorophenol ≥98%

Glass bottle for solids, 250 g

CAS Number:  88-06-2
Price
50,70 €
Log in for availability
1 * 250 g
Product Details & Documents

Purity: 98% CAS No.: 88-06-2 Molecular Formula: C6H3Cl3O Formula Weight: 197.45 Flash Point: 99°(210°F) Physical Form: Cryst solid Density: 1.675 Appearance: White to pale brown Odour: Phenol-like Melting Point: 63-68° Boiling Point: 245-246° Beilstein Reference: 776729 Merck Reference: 14,9644 UN No.: 2020 MDL No.: MFCD00002172

  • CAS Number:88-06-2
  • Formula:C₆H₃Cl₃O
  • MDL Number:MFCD00002172
  • Molecular Weight:197.45 g/mol
  • Merck Index:14,9644
  • Boiling Pt:245…246 °C
  • Melting Pt:63…68 °C
  • Density:1.675
  • Flash Pt:99 °C (210 °F)
  • Storage Temperature:Ambient
  • EINECS:201-795-9
  • UN:2020
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications