Order Entry
Export
Orders LinkContactUsLinkComponent
Bronopol ≥98%

Bronopol ≥98%

Glass bottle for solids, 500 g
Catalog # A11639.36
Toxic
Corrosive
Flammable
Irritant
Bronopol ≥98%
Catalog # A11639.36
CAS Number:  52-51-7
Bronopol ≥98%
Bronopol ≥98%

Glass bottle for solids, 500 g

CAS Number:  52-51-7
Price
506,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 98+% CAS No.: 52-51-7 Molecular Formula: C3H6BrNO4 Formula Weight: 199.99 pH: 5-7 Melting Point: ca 122-132° Beilstein Reference: 1705868 Merck Reference: 14,1447 UN No.: 3241 MDL No.: MFCD00007390

Synonyms:  2-Bromo-2-nitro-1,3-propanediol, 2-Bromo-2-nitropropane-1,3-diol
  • CAS Number:52-51-7
  • Formula:HOCH₂BrC(NO₂)CH₂OH
  • MDL Number:MFCD00007390
  • Molecular Weight:199.99 g/mol
  • Merck Index:14,1447
  • Boiling Pt:388 °C (1013 hPa)
  • Melting Pt:∼122…132 °C
  • Density:1.91 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-143-0
  • UN:3241
  • ADR:4.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications