Orders LinkContactUsLinkComponent
Bronopol ≥98%
Glass bottle for solids, 100 g
  A11639.22
Bronopol ≥98%
  A11639.22
 :  52-51-7
Bronopol ≥98%
Bronopol ≥98%
Glass bottle for solids, 100 g
 :  52-51-7
157,00 €
1 * 100 g
Purity: 98+% CAS No.: 52-51-7 Molecular Formula: C3H6BrNO4 Formula Weight: 199.99 pH: 5-7 Melting Point: ca 122-132° Beilstein Reference: 1705868 Merck Reference: 14,1447 UN No.: 3241 MDL No.: MFCD00007390
 :  2-Bromo-2-nitro-1,3-propanediol, 2-Bromo-2-nitropropane-1,3-diol
  •  :52-51-7
  •  :HOCH₂BrC(NO₂)CH₂OH
  •  :MFCD00007390
  •  :199.99 g/mol
  •  :14,1447
  •  :388 °C (1013 hPa)
  •  :∼122…132 °C
  •  :1.91 g/cm³ (20 °C)
  •  :Ambient
  •  :200-143-0
  •  :3241
  •  :4.1,III
  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g