Order Entry
Export
Orders LinkContactUsLinkComponent
 

3,4-Dihydroxybenzaldehyde ≥98%

Glass bottle for solids, 25 g
Catalog # A11558.14
Irritant
3,4-Dihydroxybenzaldehyde ≥98%
Catalog # A11558.14
CAS Number:  139-85-5
3,4-Dihydroxybenzaldehyde ≥98%
3,4-Dihydroxybenzaldehyde ≥98%

Glass bottle for solids, 25 g

CAS Number:  139-85-5
Price
166,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98% CAS No.: 139-85-5 Molecular Formula: C7H6O3 Formula Weight: 138.12 Melting Point: 152-157° Beilstein Reference: 774381 Merck Reference: 14,7893 MDL No.: MFCD00003370

Synonyms:  Protocatechuic aldehyde, Protocatechualdehyde
  • CAS Number:139-85-5
  • Formula:C₇H₆O₃
  • MDL Number:MFCD00003370
  • Molecular Weight:138.12 g/mol
  • Merck Index:14,7893
  • Boiling Pt:295 °C (1013 hPa)
  • Melting Pt:152…157 °C
  • Density:1.505 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:205-377-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications