Thymidine (2'-deoxythymidine) ≥99%
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
Thymidine (2'-deoxythymidine) ≥99%
Catalog # A11493.22
Supplier: Thermo Fisher Scientific
CAS Number: 50-89-5
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 50-89-5 Molecular Formula: C10H14N2O5 Formula Weight: 242.23 Physical Form: Powder Appearance: White Odour: Odourless Melting Point: 186-190° Beilstein Reference: 89285 Merck Reference: 14,9397 Optical Rotation: +19° (c=1 in water) MDL No.: MFCD00006537
Synonyms: 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione, 1-(2-Deoxy-β-D-ribofuranosyl)thymine, 1-(2-Deoxy-β-D-ribofuranosyl)-5-methyluracil, dT, Thymine deoxyriboside, 1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione, Pyrimidine deoxynucleoside, Thymidine kinase, Thymidine-2-deoxythymidine
- CAS Number:50-89-5
- Formula:C₁₀H₁₄N₂O₅
- MDL Number:MFCD00006537
- Molecular Weight:242.23 g/mol
- Merck Index:14,9397
- Melting Pt:186…190 °C
- Storage Temperature:Ambient
- EINECS:200-070-4
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



