Order Entry
Export
Orders LinkContactUsLinkComponent
Thymidine (2'-deoxythymidine) ≥99%

Thymidine (2'-deoxythymidine) ≥99%

Glass bottle for solids, 5 g
  A11493.06
Health
Irritant
Thymidine (2'-deoxythymidine) ≥99%
  A11493.06
 :  50-89-5
Thymidine (2'-deoxythymidine) ≥99%
Thymidine (2'-deoxythymidine) ≥99%

Glass bottle for solids, 5 g

 :  50-89-5
 
114,00 €
Log in for availability
1 * 5 g
 

Purity: 99% CAS No.: 50-89-5 Molecular Formula: C10H14N2O5 Formula Weight: 242.23 Physical Form: Powder Appearance: White Odour: Odourless Melting Point: 186-190° Beilstein Reference: 89285 Merck Reference: 14,9397 Optical Rotation: +19° (c=1 in water) MDL No.: MFCD00006537

Synonyms:  1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione, 1-(2-Deoxy-β-D-ribofuranosyl)thymine, 1-(2-Deoxy-β-D-ribofuranosyl)-5-methyluracil, dT, Thymine deoxyriboside, 1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione, Pyrimidine deoxynucleoside, Thymidine kinase, Thymidine-2-deoxythymidine
  • CAS Number:50-89-5
  • Formula:C₁₀H₁₄N₂O₅
  • MDL Number:MFCD00006537
  • Molecular Weight:242.23 g/mol
  • Merck Index:14,9397
  • Melting Pt:186…190 °C
  • Storage Temperature:Ambient
  • EINECS:200-070-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications