Order Entry
Export
Orders LinkContactUsLinkComponent
Hydroquinone ≥99%

Hydroquinone ≥99%

Glass bottle for solids, 250 g
Catalog # A11411.30
Corrosive
Health
Irritant
Environment
Hydroquinone ≥99%
Catalog # A11411.30
CAS Number:  123-31-9
Hydroquinone ≥99%
Hydroquinone ≥99%

Glass bottle for solids, 250 g

CAS Number:  123-31-9
Price
44,60 €
Log in for availability
1 * 250 g
Product Details & Documents

Purity: 99% CAS No.: 123-31-9 Molecular Formula: C6H6O2 Formula Weight: 110.11 Flash Point: 165°(329°F) Odour: Odourless pH: 3.75 Melting Point: 170-172° Boiling Point: 285-287° Beilstein Reference: 605970 Merck Reference: 14,4808 UN No.: 3077 MDL No.: MFCD00002339

Synonyms:  1,4-Dihydroxybenzene, Quinol
  • CAS Number:123-31-9
  • Formula:C₆H₄(OH)₂
  • MDL Number:MFCD00002339
  • Molecular Weight:110.11 g/mol
  • Merck Index:14,4808
  • Boiling Pt:285…287 °C
  • Melting Pt:170…172 °C
  • Density:1.320
  • Flash Pt:165 °C (329 °F)
  • Storage Temperature:Ambient
  • EINECS:204-617-8
  • UN:3077
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications