2,6-Dichloropyridine ≥98%
Glass bottle for solids, 2.500 g
Supplier: Thermo Fisher Scientific
2,6-Dichloropyridine ≥98%
Catalog # A11364.0E
Supplier: Thermo Fisher Scientific
CAS Number: 2402-78-0
Jump to:
Product Details & Documents
Purity: 98% CAS No.: 2402-78-0 Molecular Formula: C5H3Cl2N Formula Weight: 147.99 Odour: Unpleasant Melting Point: 84-88° Boiling Point: 211° Beilstein Reference: 108664 UN No.: 2811 MDL No.: MFCD00006244
- CAS Number:2402-78-0
- Formula:C₅H₃Cl₂N
- MDL Number:MFCD00006244
- Molecular Weight:147.99 g/mol
- Boiling Pt:211 °C
- Melting Pt:84…88 °C
- Storage Temperature:Ambient
- EINECS:219-282-3
- UN:2811
- ADR:6.1,III
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:2.500 g



