D-(-)-Serine ≥99%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 312-84-5 Molecular Formula: C3H7NO3 Formula Weight: 105.09 Melting Point: 218-220° dec. Beilstein Reference: 1721403 Merck Reference: 14,8460 Optical Rotation: -13° (c=5 in 5N HCl) MDL No.: MFCD00004269
Synonyms: (R)-(-)-Serine, H-D-Ser-OH
- CAS Number:312-84-5
- Formula:C₃H₇NO₃
- MDL Number:MFCD00004269
- Molecular Weight:105.09 g/mol
- Merck Index:14,8460
- Melting Pt:218…220 °C (dec.)
- Storage Temperature:Ambient
- EINECS:206-229-4
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



