Order Entry
Export
Orders LinkContactUsLinkComponent
3,4-Dimethoxyphenol ≥98%

3,4-Dimethoxyphenol ≥98%

Glass bottle for solids, 5 g
Catalog # A11344.06
Irritant
3,4-Dimethoxyphenol ≥98%
Catalog # A11344.06
CAS Number:  2033-89-8
3,4-Dimethoxyphenol ≥98%
3,4-Dimethoxyphenol ≥98%

Glass bottle for solids, 5 g

CAS Number:  2033-89-8
Price
95,10 €
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 98%
CAS No.: 2033-89-8
Molecular Formula: C8H10O3
Formula Weight: 154.16
Melting Point: 79-82°
Boiling Point: 167°/14mm
MDL No.: MFCD00008390

  • CAS Number:2033-89-8
  • Formula:C₈H₁₀O₃
  • MDL Number:MFCD00008390
  • Molecular Weight:154.17 g/mol
  • Boiling Pt:167 °C (14 mmHg)
  • Melting Pt:79…82 °C
  • Storage Temperature:Ambient
  • EINECS:217-995-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications