Orders LinkContactUsLinkComponent
1,3-Diethoxybenzene 95%
Glass bottle for solids, 50 g
  A11338.18
1,3-Diethoxybenzene 95%
  A11338.18
 :  2049-73-2
1,3-Diethoxybenzene 95%
1,3-Diethoxybenzene 95%
Glass bottle for solids, 50 g
 :  2049-73-2
103,00 €
1 * 50 g
Purity: 95% CAS No.: 2049-73-2 Molecular Formula: C10H14O2 Formula Weight: 166.22 Refractive Index: 1.5095 Melting Point: 11-12° Boiling Point: 234-236° Beilstein Reference: 2517730 MDL No.: MFCD00015145
  •  :2049-73-2
  •  :C₁₀H₁₄O₂
  •  :MFCD00015145
  •  :166.22 g/mol
  •  :234…236 °C
  •  :11…12 °C
  •  :1.010
  •  :Ambient
  •  :218-071-3
  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g