Paracetamol ≥98%
Glass bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Acetaminophen, N-Acetyl-4-aminophenol, 4′-Hydroxyacetanilide, 4'-Hydroxyacetanilide, Paracetamolum, 4-Acetamidophenol, N-(4-Hydroxyphenyl)acetamide, APAP
- CAS Number:103-90-2
- Formula:C₈H₉NO₂
- MDL Number:MFCD00002328
- Molecular Weight:151.16 g/mol
- Merck Index:14,47
- Boiling Pt:388 °C (1013 hPa)
- Melting Pt:168…172 °C
- Density:1.293
- Storage Temperature:Ambient
- EINECS:203-157-5
Documents
Specifications
Specification Test Results
- Appearance (Color):White
- Form:crystalline powder
- Assay (HPLC):97.5%
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Cat. No.:A11240.36



