Paracetamol ≥98%
Glass bottle for solids, 250 g
: Acetaminophen, N-Acetyl-4-aminophenol, 4′-Hydroxyacetanilide, 4'-Hydroxyacetanilide, Paracetamolum, 4-Acetamidophenol, N-(4-Hydroxyphenyl)acetamide, APAP
- :103-90-2
- :C₈H₉NO₂
- :MFCD00002328
- :151.16 g/mol
- :14,47
- :388 °C (1013 hPa)
- :168…172 °C
- :1.293
- :Ambient
- :203-157-5
- Appearance (Color):White
- Form:crystalline powder
- Assay (HPLC):97.5%
- Pack type:Glass bottle for solids
- Pk:250 g



