Order Entry
Export
Orders LinkContactUsLinkComponent
2,6-Dichloropyrazine ≥98%

2,6-Dichloropyrazine ≥98%

Glass bottle for solids, 25 g
Catalog # A11175.14
Irritant
2,6-Dichloropyrazine ≥98%
Catalog # A11175.14
CAS Number:  4774-14-5
2,6-Dichloropyrazine ≥98%
2,6-Dichloropyrazine ≥98%

Glass bottle for solids, 25 g

CAS Number:  4774-14-5
Price
548,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 99% CAS No.: 4774-14-5 Molecular Formula: C4H2Cl2N2 Formula Weight: 148.98 Flash Point: 110°(230°F) Melting Point: 52-56° Beilstein Reference: 2439 MDL No.: MFCD00006125

  • CAS Number:4774-14-5
  • Formula:C₄H₂Cl₂N₂
  • MDL Number:MFCD00006125
  • Molecular Weight:148.98 g/mol
  • Boiling Pt:120…122 °C (40 mmHg)
  • Melting Pt:52…56 °C
  • Density:1.713 g/cm³ (20 °C)
  • Flash Pt:>110 °C (230 °F)
  • Storage Temperature:Ambient
  • EINECS:225-316-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications