Orders LinkContactUsLinkComponent
4-Hydroxy-2-methoxybenzaldehyde ≥98%
4-Hydroxy-2-methoxybenzaldehyde ≥98%
Glass bottle for solids, 25 g
  A11127.14
4-Hydroxy-2-methoxybenzaldehyde ≥98%
  A11127.14
 :  18278-34-7
4-Hydroxy-2-methoxybenzaldehyde ≥98%
4-Hydroxy-2-methoxybenzaldehyde ≥98%
Glass bottle for solids, 25 g
 :  18278-34-7
490,00 €
1 * 25 g
Purity: 98% CAS No.: 18278-34-7 Molecular Formula: C8H8O3 Formula Weight: 152.15 Melting Point: 156-160° Beilstein Reference: 2045122
  •  :18278-34-7
  •  :C₈H₈O₃
  •  :MFCD00051964
  •  :152.15 g/mol
  •  :273 °C (1013 hPa)
  •  :156…160 °C
  •  :Ambient
  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g