Order Entry
Export
Orders LinkContactUsLinkComponent
4-Hydroxy-2-methoxybenzaldehyde ≥98%

4-Hydroxy-2-methoxybenzaldehyde ≥98%

Glass bottle for solids, 5 g
Catalog # A11127.06
Irritant
4-Hydroxy-2-methoxybenzaldehyde ≥98%
Catalog # A11127.06
CAS Number:  18278-34-7
4-Hydroxy-2-methoxybenzaldehyde ≥98%
4-Hydroxy-2-methoxybenzaldehyde ≥98%

Glass bottle for solids, 5 g

CAS Number:  18278-34-7
Price
130,00 €
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 98% CAS No.: 18278-34-7 Molecular Formula: C8H8O3 Formula Weight: 152.15 Melting Point: 156-160° Beilstein Reference: 2045122

  • CAS Number:18278-34-7
  • Formula:C₈H₈O₃
  • MDL Number:MFCD00051964
  • Molecular Weight:152.15 g/mol
  • Boiling Pt:273 °C (1013 hPa)
  • Melting Pt:156…160 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications