Order Entry
Export
Orders LinkContactUsLinkComponent
 

2-Nitrobenzonitrile ≥98%

Glass bottle for solids, 25 g
Catalog # A10941.14
Toxic
2-Nitrobenzonitrile ≥98%
Catalog # A10941.14
CAS Number:  612-24-8
2-Nitrobenzonitrile ≥98%
2-Nitrobenzonitrile ≥98%

Glass bottle for solids, 25 g

CAS Number:  612-24-8
Price
107,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98+% CAS No.: 612-24-8 Molecular Formula: C7H4N2O2 Formula Weight: 148.12 Odour: Almond-like Melting Point: 107-111° Boiling Point: 165°/16mm Beilstein Reference: 638987 UN No.: 3439 MDL No.: MFCD00007044

Synonyms:  o-Nitrobenzonitrile
  • CAS Number:612-24-8
  • Formula:C₇H₄N₂O₂
  • MDL Number:MFCD00007044
  • Molecular Weight:148.12 g/mol
  • Boiling Pt:165 °C (16 mmHg)
  • Melting Pt:107…111 °C
  • Storage Temperature:Ambient
  • EINECS:210-301-0
  • UN:3439
  • ADR:6.1,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications