Barbituric acid ≥99%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 67-52-7 Molecular Formula: C4H4N2O3 Formula Weight: 128.09 Flash Point: 150°(302°F) pH: 2-3 Melting Point: ca 250° dec. Beilstein Reference: 120502 Merck Reference: 14,963 MDL No.: MFCD00006666
Synonyms: N,N'-Malonylurea, 2,4,6-Trihydroxypyrimidine
- CAS Number:67-52-7
- Formula:C₄H₄N₂O₃
- MDL Number:MFCD00006666
- Molecular Weight:128.09 g/mol
- Merck Index:14,963
- Boiling Pt:441 °C (1013 hPa)
- Melting Pt:∼ 250 °C (dec.)
- Flash Pt:150 °C (302 °F)
- Storage Temperature:Ambient
- EINECS:200-658-0
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



