Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,3-Dimethylaniline ≥98%

Glass bottle for solids, 2.500 g
Catalog # A10918.0E
Toxic
Health
Environment
2,3-Dimethylaniline ≥98%
Catalog # A10918.0E
CAS Number:  87-59-2
2,3-Dimethylaniline ≥98%
2,3-Dimethylaniline ≥98%

Glass bottle for solids, 2.500 g

CAS Number:  87-59-2
Price
381,00 €
Log in for availability
1 * 2.500 g
Product Details & Documents

Purity: 98% CAS No.: 87-59-2 Molecular Formula: C8H11N Formula Weight: 121.18 Flash Point: 96°(204°F) Refractive Index: 1.5680 Odour: Characteristic Melting Point: 2-3° Boiling Point: 221-222° Beilstein Reference: 742174 Merck Reference: 14,10084 UN No.: 1711 MDL No.: MFCD00007732

Synonyms:  2,3-Xylidine, 3-Amino-o-xylene
  • CAS Number:87-59-2
  • Formula:C₈H₁₁N
  • MDL Number:MFCD00007732
  • Molecular Weight:121.18 g/mol
  • Merck Index:14,10084
  • Boiling Pt:221…222 °C
  • Melting Pt:2…3 °C
  • Density:0.992
  • Flash Pt:96 °C (204 °F)
  • Storage Temperature:Ambient
  • EINECS:201-755-0
  • UN:1711
  • ADR:6.1,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2.500 g

Specifications