Order Entry
Export
Orders LinkContactUsLinkComponent
(4-Bromophenyl)acetonitrile ≥97%

(4-Bromophenyl)acetonitrile ≥97%

Glass bottle for solids, 500 g
Catalog # A10887.36
Toxic
Irritant
(4-Bromophenyl)acetonitrile ≥97%
Catalog # A10887.36
CAS Number:  16532-79-9
(4-Bromophenyl)acetonitrile ≥97%
(4-Bromophenyl)acetonitrile ≥97%

Glass bottle for solids, 500 g

CAS Number:  16532-79-9
Price
450,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 97% CAS No.: 16532-79-9 Molecular Formula: C8H6BrN Formula Weight: 196.05 Flash Point: 127°(261°F) Odour: Pungent Boiling Point: 286° Beilstein Reference: 1861069 UN No.: 3449 MDL No.: MFCD00001916

Synonyms:  4-Bromophenylacetonitrile, 4-Bromobenzyl cyanide
  • CAS Number:16532-79-9
  • Formula:C₈H₆BrN
  • MDL Number:MFCD00001916
  • Molecular Weight:196.05 g/mol
  • Boiling Pt:286 °C
  • Melting Pt:45…49 °C
  • Density:1.49
  • Flash Pt:127 °C (261 °F)
  • Storage Temperature:Ambient
  • EINECS:240-602-2
  • UN:3449
  • ADR:6.1,I
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications