Orders LinkContactUsLinkComponent
Diphenylsilane ≥97%
Glass bottle for solids, 5 g
  A10884.06
Diphenylsilane ≥97%
  A10884.06
 :  775-12-2
Diphenylsilane ≥97%
Diphenylsilane ≥97%
Glass bottle for solids, 5 g
 :  775-12-2
54,80 €
1 * 5 g
Purity: 97% CAS No.: 775-12-2 Molecular Formula: C12H12Si Formula Weight: 184.32 Flash Point: 98°(208°F) Refractive Index: 1.5790 Boiling Point: 95-97°/13mm Beilstein Reference: 2935887 MDL No.: MFCD00003002
  •  :775-12-2
  •  :C₁₂H₁₂Si
  •  :MFCD00003002
  •  :184.31 g/mol
  •  :95…97 °C (13 mmHg)
  •  :0.996
  •  :98 °C (208 °F)
  •  :Ambient
  •  :212-271-4
  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g