Order Entry
Export
Orders LinkContactUsLinkComponent
 

ω,2,4-Trichloroacetophenone ≥97%

Glass bottle for solids, 100 g
Catalog # A10835.22
Toxic
Corrosive
Irritant
ω,2,4-Trichloroacetophenone ≥97%
Catalog # A10835.22
CAS Number:  4252-78-2
ω,2,4-Trichloroacetophenone ≥97%
ω,2,4-Trichloroacetophenone ≥97%

Glass bottle for solids, 100 g

CAS Number:  4252-78-2
Price
77,70 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 97% CAS No.: 4252-78-2 Molecular Formula: C8H5Cl3O Formula Weight: 223.49 Flash Point: 110°(230°F) Melting Point: 52-56° Beilstein Reference: 957098 UN No.: 2923 MDL No.: MFCD00000934

Synonyms:  2,4-Dichlorophenacyl chloride, α,2,4-Trichloroacetophenone, 2,2',4'-Trichloroacetophenone
  • CAS Number:4252-78-2
  • Formula:C₈H₅Cl₃O
  • MDL Number:MFCD00000934
  • Molecular Weight:223.49 g/mol
  • Boiling Pt:292 °C (1013 hPa)
  • Melting Pt:52…56 °C
  • Flash Pt:>110 °C (230 °F)
  • Storage Temperature:Ambient
  • EINECS:224-218-2
  • UN:2923
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications