Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,3-Propanediol ≥99%

Glass bottle for solids, 250 g
Catalog # A10829.30
   Safer Human and Environmental Health
1,3-Propanediol ≥99%
Catalog # A10829.30
   Safer Human and Environmental Health
CAS Number:  504-63-2
1,3-Propanediol ≥99%
1,3-Propanediol ≥99%

Glass bottle for solids, 250 g

CAS Number:  504-63-2
Price
68,50 €
Log in for availability
1 * 250 g
Product Details & Documents

Purity: 99% CAS No.: 504-63-2 Molecular Formula: HO(CH2)3OH Formula Weight: 76.10 Flash Point: 140°(174°F) Refractive Index: 1.4400 Melting Point: -26° Boiling Point: 214° Solubility: Miscible with water, alcohol Beilstein Reference: 969155 Merck Reference: 14,9714 MDL No.: MFCD00002949

Synonyms:  1,3-Propylene glycol, 1,3-Dihydroxy propane
  • CAS Number:504-63-2
  • Formula:HO(CH₂)₃OH
  • MDL Number:MFCD00002949
  • Molecular Weight:76.1 g/mol
  • Merck Index:14,9714
  • Boiling Pt:214 °C
  • Melting Pt:–26 °C
  • Density:1.053
  • Flash Pt:140 °C (174 °F)
  • Storage Temperature:Ambient
  • EINECS:207-997-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g
  • Cat. No.:
    A10829.30
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating)
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications