Orders LinkContactUsLinkComponent
Adenosine ≥99%
Glass bottle for solids, 250 g
  A10781.30
Adenosine ≥99%
  A10781.30
 :  58-61-7
Adenosine ≥99%
Adenosine ≥99%
Glass bottle for solids, 250 g
 :  58-61-7
603,00 €
1 * 250 g
Purity: 99% CAS No.: 58-61-7 Molecular Formula: C10H13N5O4 Formula Weight: 267.25 Storage Temperature: 2°C to 8°C Physical Form: Powder Odour: Odourless Melting Point: 234-237° Beilstein Reference: 93029 Merck Reference: 14,153 Optical Rotation: -70° (c=2 in 5% NaOH) MDL No.: MFCD00005752
 :  9-β-D-Ribofuranosyladenine, (2R,3R,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, Adenine-9-β-D-ribofuranoside, Adenine riboside
  •  :58-61-7
  •  :C₁₀H₁₃N₅O₄
  •  :MFCD00005752
  •  :267.24 g/mol
  •  :14,153
  •  :234…237 °C
  •  :Refrigerator
  •  :200-389-9
  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g