Order Entry
Export
Orders LinkContactUsLinkComponent
Adenosine ≥99%

Adenosine ≥99%

Glass bottle for solids, 50 g
Catalog # A10781.18
Adenosine ≥99%
Catalog # A10781.18
CAS Number:  58-61-7
Adenosine ≥99%
Adenosine ≥99%

Glass bottle for solids, 50 g

CAS Number:  58-61-7
Price
154,00 €
Log in for availability
1 * 50 g
Product Details & Documents

Purity: 99% CAS No.: 58-61-7 Molecular Formula: C10H13N5O4 Formula Weight: 267.25 Storage Temperature: 2°C to 8°C Physical Form: Powder Odour: Odourless Melting Point: 234-237° Beilstein Reference: 93029 Merck Reference: 14,153 Optical Rotation: -70° (c=2 in 5% NaOH) MDL No.: MFCD00005752

Synonyms:  9-β-D-Ribofuranosyladenine, (2R,3R,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, Adenine-9-β-D-ribofuranoside, Adenine riboside
  • CAS Number:58-61-7
  • Formula:C₁₀H₁₃N₅O₄
  • MDL Number:MFCD00005752
  • Molecular Weight:267.24 g/mol
  • Merck Index:14,153
  • Melting Pt:234…237 °C
  • Storage Temperature:Refrigerator
  • EINECS:200-389-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g

Specifications