Agar, powder
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 9002-18-0 Molecular Formula: (C12H18O9)X Physical Form: Powder Appearance: White to cream Odour: Odourless Merck Reference: 14,184 MDL No.: MFCD00081288
Synonyms: Agar Agar
- CAS Number:9002-18-0
- MDL Number:MFCD00081288
- Merck Index:14,184
- Melting Pt:90 °C
- Storage Temperature:Ambient
- EINECS:232-658-1
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



