Order Entry
Export
Orders LinkContactUsLinkComponent
 

Agar, powder

Glass bottle for solids, 2.500 g
Catalog # A10752.0E
Agar, powder
Catalog # A10752.0E
CAS Number:  9002-18-0
Agar, powder
Agar, powder

Glass bottle for solids, 2.500 g

CAS Number:  9002-18-0
Price
693,00 €
Log in for availability
1 * 2.500 g
Product Details & Documents

CAS No.: 9002-18-0 Molecular Formula: (C12H18O9)X Physical Form: Powder Appearance: White to cream Odour: Odourless Merck Reference: 14,184 MDL No.: MFCD00081288

Synonyms:  Agar Agar
  • CAS Number:9002-18-0
  • MDL Number:MFCD00081288
  • Merck Index:14,184
  • Melting Pt:90 °C
  • Storage Temperature:Ambient
  • EINECS:232-658-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2.500 g

Specifications