Order Entry
Export
Orders LinkContactUsLinkComponent
 

D-(+)-Xylose ≥98%

Glass bottle for solids, 100 g
Catalog # A10643.22
D-(+)-Xylose ≥98%
Catalog # A10643.22
CAS Number:  58-86-6
D-(+)-Xylose ≥98%
D-(+)-Xylose ≥98%

Glass bottle for solids, 100 g

CAS Number:  58-86-6
Price
59,40 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 98+% CAS No.: 58-86-6 Molecular Formula: C5H10O5 Formula Weight: 150.13 Flash Point: 100°(212°F) Physical Form: Cryst powder Density: 1.525 Sensitivity: Hygroscopic Appearance: White Melting Point: 147-151° Beilstein Reference: 1562108 Merck Reference: 14,10087 Optical Rotation: +19° (c=10 in water + NH4OH) MDL No.: MFCD00151475

  • CAS Number:58-86-6
  • Formula:C₅H₁₀O₅
  • MDL Number:MFCD00151475
  • Molecular Weight:150.13 g/mol
  • Merck Index:14,10087
  • Boiling Pt:331 °C (1013 hPa)
  • Melting Pt:147…151 °C
  • Density:1.525
  • Flash Pt:>100 °C (212 °F)
  • Storage Temperature:Ambient
  • EINECS:200-400-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications