D-(+)-Xylose ≥98%
Glass bottle for solids, 2.500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 98+% CAS No.: 58-86-6 Molecular Formula: C5H10O5 Formula Weight: 150.13 Flash Point: 100°(212°F) Physical Form: Cryst powder Density: 1.525 Sensitivity: Hygroscopic Appearance: White Melting Point: 147-151° Beilstein Reference: 1562108 Merck Reference: 14,10087 Optical Rotation: +19° (c=10 in water + NH4OH) MDL No.: MFCD00151475
- CAS Number:58-86-6
- Formula:C₅H₁₀O₅
- MDL Number:MFCD00151475
- Molecular Weight:150.13 g/mol
- Merck Index:14,10087
- Boiling Pt:331 °C (1013 hPa)
- Melting Pt:147…151 °C
- Density:1.525
- Flash Pt:>100 °C (212 °F)
- Storage Temperature:Ambient
- EINECS:200-400-7
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:2.500 g



