Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,3-Dithiane ≥98%

Glass bottle for solids, 5 g
Catalog # A10505.06
1,3-Dithiane ≥98%
Catalog # A10505.06
CAS Number:  505-23-7
1,3-Dithiane ≥98%
1,3-Dithiane ≥98%

Glass bottle for solids, 5 g

CAS Number:  505-23-7
Price
57,30 €
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 98% CAS No.: 505-23-7 Molecular Formula: C4H8S2 Formula Weight: 120.24 Flash Point: 90°(194°F) Odour: Stench Boiling Point: 207-208° Beilstein Reference: 102534 UN No.: 3335 MDL No.: MFCD00006654

  • CAS Number:505-23-7
  • Formula:C₄H₈S₂
  • MDL Number:MFCD00006654
  • Molecular Weight:120.24 g/mol
  • Boiling Pt:207…208 °C
  • Melting Pt:52…55 °C
  • Density:1.36 g/cm³ (25 °C)
  • Flash Pt:90 °C (194 °F)
  • Storage Temperature:Ambient
  • EINECS:208-006-7
  • ADR:9,III
  •  :Density: 1.36 g/cm³ (25 °C)
  • UN:3335
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications