Glutaraldehyde 50% in aqueous solution
Glass bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
Glutaraldehyde 50% in aqueous solution
Catalog # A10500.36
Supplier: Thermo Fisher Scientific
CAS Number: 111-30-8
Jump to:
Product Details & Documents
CAS No.: 111-30-8 Molecular Formula: C5H8O2 Formula Weight: 100.12 Storage Temperature: 2°C to 8°C Physical Form: Liquid Density: 1.123 Refractive Index: 1.4210 Appearance: Colourless to pale yellow Odour: Stench Melting Point: ca -21° Boiling Point: 100° Beilstein Reference: 605390 Merck Reference: 14,4472 UN No.: 2922 MDL No.: MFCD00007025
Synonyms: Glutaral, Glutardialdehyde, 1,5-Pentanedial
- CAS Number:111-30-8
- Formula:C₅H₈O₂
- MDL Number:MFCD00007025
- Molecular Weight:100.12 g/mol
- Merck Index:14,4472
- Boiling Pt:100 °C
- Melting Pt:∼–21 °C
- Density:1.123
- Storage Temperature:Refrigerator
- EINECS:203-856-5
- ADR:8,III
- :Density: 1.014 g/cm³ (20 °C)
- UN:2922
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g



