Order Entry
Export
Orders LinkContactUsLinkComponent
Glutaraldehyde 50% in aqueous solution

Glutaraldehyde 50% in aqueous solution

Glass bottle for solids, 100 g
Catalog # A10500.22
Toxic
Health
Corrosive
Environment
Glutaraldehyde 50% in aqueous solution
Catalog # A10500.22
CAS Number:  111-30-8
Glutaraldehyde 50% in aqueous solution
Glutaraldehyde 50% in aqueous solution

Glass bottle for solids, 100 g

CAS Number:  111-30-8
Price
27,30 €
Log in for availability
1 * 100 g
Product Details & Documents

CAS No.: 111-30-8 Molecular Formula: C5H8O2 Formula Weight: 100.12 Storage Temperature: 2°C to 8°C Physical Form: Liquid Density: 1.123 Refractive Index: 1.4210 Appearance: Colourless to pale yellow Odour: Stench Melting Point: ca -21° Boiling Point: 100° Beilstein Reference: 605390 Merck Reference: 14,4472 UN No.: 2922 MDL No.: MFCD00007025

Synonyms:  Glutaral, Glutardialdehyde, 1,5-Pentanedial
  • CAS Number:111-30-8
  • Formula:C₅H₈O₂
  • MDL Number:MFCD00007025
  • Molecular Weight:100.12 g/mol
  • Merck Index:14,4472
  • Boiling Pt:100 °C
  • Melting Pt:∼–21 °C
  • Density:1.123
  • Storage Temperature:Refrigerator
  • EINECS:203-856-5
  • ADR:8,III
  •  :Density: 1.014 g/cm³ (20 °C)
  • UN:2922
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications