Order Entry
Export
Orders LinkContactUsLinkComponent
1,12-Dodecanedioic acid ≥99%

1,12-Dodecanedioic acid ≥99%

Glass bottle for solids, 250 g
Catalog # A10387.30
Irritant
1,12-Dodecanedioic acid ≥99%
Catalog # A10387.30
CAS Number:  693-23-2
1,12-Dodecanedioic acid ≥99%
1,12-Dodecanedioic acid ≥99%

Glass bottle for solids, 250 g

CAS Number:  693-23-2
Price
46,90 €
Log in for availability
1 * 250 g
Product Details & Documents

Purity: 99% CAS No.: 693-23-2 Molecular Formula: HO2C(CH2)10CO2H Formula Weight: 230.31 Flash Point: 227°(440°F) Odour: Odourless Boiling Point: 244-245°/10mm Beilstein Reference: 1782580 MDL No.: MFCD00002735

Synonyms:  Dodecanedioic acid, 1,10-Decanedicarboxylic acid
  • CAS Number:693-23-2
  • Formula:C₁₂H₂₂O₄
  • MDL Number:MFCD00002735
  • Molecular Weight:230.3 g/mol
  • Boiling Pt:244…245 °C (10 mmHg)
  • Melting Pt:127…131 °C
  • Density:1.150
  • Flash Pt:227 °C (440 °F)
  • Storage Temperature:Ambient
  • EINECS:211-746-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications