n-Hexadecane ≥99%
Product Details & Documents
Hexadecane is use in the recovery of valuable amines by means of distillation from aqueous alkanolamine solutions, which are extensively used in the gas-processing industries.
Purity: 99% CAS No.: 544-76-3 Molecular Formula: CH3(CH2)14CH3 Formula Weight: 226.45 Flash Point: 135°(275°F) Physical Form: Liquid Density: 0.773 Refractive Index: 1.4345 Appearance: Colourless Melting Point: 18° Boiling Point: 287° Solubility: This product is soluble in acetone, alcohol, ether. It is insoluble in water. Beilstein Reference: 1736592 MDL No.: MFCD00008998
Hexadecane is use in the recovery of valuable amines by means of distillation from aqueous alkanolamine solutions, which are extensively used in the gas-processing industries. *Majid Abedinzadegan Abdi and Axel Meisen. Mutual Solubility of Hexadecane with Diethanolamine and with Bis(hydroxyethyl)piperazine J. Chem. Eng. Data, 1998, 43 (2), pp 138-142
- CAS Number:544-76-3
- Formula:H₃C(CH₂)₁₄CH₃
- MDL Number:MFCD00008998
- Molecular Weight:226.45 g/mol
- Boiling Pt:287 °C
- Melting Pt:18 °C
- Density:0.773
- Flash Pt:135 °C (275 °F)
- Storage Temperature:Ambient
- EINECS:208-878-9
Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:100 mL



