D-Alanine ≥99%
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 338-69-2 Molecular Formula: C3H7NO2 Formula Weight: 89.09 Melting Point: ca 295° dec. Beilstein Reference: 1720249 Merck Reference: 14,204 Optical Rotation: -14° (c=5 in 5N HCl) MDL No.: MFCD00008077
Synonyms: (R)-(-)-Alanine, Alanine, D-α-Alanine, D(-)-Alanine, H-D-Ala-OH, α-Alanine
- CAS Number:338-69-2
- Formula:C₃H₇NO₂
- MDL Number:MFCD00008077
- Molecular Weight:89.09 g/mol
- Merck Index:14,204
- Boiling Pt:189 °C (1013 hPa)
- Melting Pt:∼ 295 °C (dec.)
- Storage Temperature:Ambient
- EINECS:206-418-1
Documents
Certificates
Search by Lot NumberALFAA10231.14_TDS_EN
Product specification
Organic Building Blocks and Intermediates
Application Notes
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



