Order Entry
Export
Orders LinkContactUsLinkComponent
D-Alanine ≥99%

D-Alanine ≥99%

Glass bottle for solids, 5 g
Catalog # A10231.06
D-Alanine ≥99%
Catalog # A10231.06
CAS Number:  338-69-2
D-Alanine ≥99%
D-Alanine ≥99%

Glass bottle for solids, 5 g

CAS Number:  338-69-2
Price
49,70 €
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 99% CAS No.: 338-69-2 Molecular Formula: C3H7NO2 Formula Weight: 89.09 Melting Point: ca 295° dec. Beilstein Reference: 1720249 Merck Reference: 14,204 Optical Rotation: -14° (c=5 in 5N HCl) MDL No.: MFCD00008077

Synonyms:  (R)-(-)-Alanine, Alanine, D-α-Alanine, D(-)-Alanine, H-D-Ala-OH, α-Alanine
  • CAS Number:338-69-2
  • Formula:C₃H₇NO₂
  • MDL Number:MFCD00008077
  • Molecular Weight:89.09 g/mol
  • Merck Index:14,204
  • Boiling Pt:189 °C (1013 hPa)
  • Melting Pt:∼ 295 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:206-418-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications