Imidazole ≥99%
Glass bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 288-32-4 Molecular Formula: C3H4N2 Formula Weight: 68.08 Flash Point: 145°(293°F) Physical Form: Cryst flakes Density: 1.03 Appearance: White to cream Melting Point: 88-92° Boiling Point: 256° Beilstein Reference: 103853 Merck Reference: 14,4912 UN No.: 2923 MDL No.: MFCD00005183
Excellent for buffer use in pH range 6.2 - 7.8. Also an histamine antagonist.
Synonyms: Glyoxaline, 1H-Imidazole, Glyoxalamine
- CAS Number:288-32-4
- Formula:C₃H₄N₂
- MDL Number:MFCD00005183
- Molecular Weight:68.08 g/mol
- Merck Index:14,4912
- Boiling Pt:256 °C
- Melting Pt:88…92 °C
- Density:1.03
- Flash Pt:145 °C (293 °F)
- Storage Temperature:Ambient
- EINECS:206-019-2
- UN:3263
- ADR:8,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g



